Learn More
Bio X Cell InVivoMAb Isotype Control, Rat IgG2b, Anti-Keyhole Limpet Hemocyanin, Unconjugated
Supplier: Bio X Cell BE0090R025MG
The LTF-2 monoclonal antibody reacts with keyhole limpet hemocyanin (KLH). Since KLH is not expressed by mammals, this antibody is suitable as an isotype-matched control for rat IgG2b antibodies in most in vivo and in vitro applications.
- Monoclonal antibody, clone LTF-2
- Host species is rat
- Isotype IgG2b, κ
- Targets keyhole limpet hemocyanin (KLH)
- Unconjugated format
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin levels ≤1 EU/mg
- Purity ≥95% (SDS-PAGE)
- Sterilized by 0.2 µm filtration
- Produced from cell culture supernatant in an animal-free facility
- Purified by protein G
- Molecular weight of 150 kDa
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.