Learn More
Medchemexpress LLC Isovaleramide (3-methylbutanamide) | 541-46-8 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB122950MG
Encompass_Preferred
Isovaleramide (3-Methylbutanamide) is an orally active anticonvulsant. It inhibits alcohol dehydrogenase (ADH) activity and regulates the GABAergic system. Isovaleramide reduces acute kidney injury and has antiepileptic, anxiolytic, sedative, and hypnotic effects. It exhibits a 90% index protection against maximal electroshock seizure in mice and can reduce the mortality rate and attenuate pathological damage of kidney tissues in EG-poisoned rats by down-regulating the expression of serum biomarkers of AKI.
  • Orally active anticonvulsant.
  • Inhibits alcohol dehydrogenase (ADH) activity.
  • Regulates the GABAergic system.
  • Reduces acute kidney injury.
  • Possesses antiepileptic, anxiolytic, sedative, and hypnotic effects.
  • Exhibits a 90% index protection against maximal electroshock seizure in mice.
Catalog No. 50-004-11835