Learn More
Cayman Chemical IsovaleroylglycIn 500mg
SDP
Supplier:  Cayman Chemical 17581500 MG
Encompass_Preferred
Urinary metabolite of leucine that is used as a diagnostic tool in the diagnosis of various acidemias and mitochondrial fatty acid β-oxidation defects
Catalog No. 50-296-8051