Learn More
Medchemexpress LLC Isovanillic acid | 645-08-9 | 10 MG
Supplier: Medchemexpress LLC HYN686410MG
Isovanillic acid, also known as 3-Hydroxy-4-methoxybenzoic acid, is a phenolic acid naturally isolated from Scrophularia ningpoensis. It exhibits anti-thrombotic activity and is used as a laboratory chemical and for the manufacture of substances.
- Phenolic acid isolated from Scrophularia ningpoensis
- Exhibits anti-thrombotic activity
- Suitable for laboratory research
- Used in the manufacture of substances