Learn More
TARGETMOL CHEMICALS INC Isoviolanthin 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T137412MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Isoviolanthin reduces the migratory and invasive capacities of TGF-beta1-treated HCC cells without cytotoxic effects on normal live cells and has potential as a therapeutic agent for the treatment of advanced-stage metastatic HCC. Isoviolanthin is a flavonoid glycoside extracted from the leaves of Dendrobium Officinale. Purity 99.61%
Catalog No. 50-288-0446