Learn More
Medchemexpress LLC Isoxsuprine-d6 hydrochloride | 2706004-35-3 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1270S1MG
Encompass_Preferred
Isoxsuprine-d6 hydrochloride is a deuterium-labeled version of Isoxsuprine hydrochloride, a beta-adrenergic receptor agonist and NMDA receptor antagonist. Stable heavy isotopes such as deuterium are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12837