Learn More
Selleck Chemical LLC Ispinesib (SB-715992) 10mg 336113-53-2 CK0238273
SDP
Supplier:  Selleck Chemical LLC S145210MG
Encompass_Preferred
Ispinesib (SB-715992, CK0238273) is a potent, specific and reversible inhibitor of kinesin spindle protein (KSP) with Ki app of 1.7 nM in a cell-free assay, no inhibition to CENP-E, RabK6, MCAK, MKLP1, KHC or Kif1A. Ispinesib induces mitotic arrest and apoptotic cell death. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-857-7