Learn More
Medchemexpress LLC Isradipine | 75695-93-1 | MFCD00153820 | >98.0% | 371.39 | C19H21N3O5 | 10 MG

Supplier: Medchemexpress LLC HYB023310MG
Isradipine is a dihydropyridine L-type calcium channel blocker used in research to modulate Cav1 family channels and study calcium-dependent cellular processes. Supplied as a research-grade solid in small pack sizes, it has molecular formula C19H21N3O5 and molecular weight 371.39 g/mol. Purity is specified per lot on the Certificate of Analysis; commercial grades are commonly ≥98% (HPLC).
- Dihydropyridine L-type calcium channel blocker for research use.
- Suitable for studies of calcium-dependent signaling and neuroprotection.
- Available in small pack sizes including 10 mg for screening and assay work.
- Molecular formula C19H21N3O5, molecular weight 371.39 g/mol.
- CAS number 75695-93-1 for unambiguous identification.
- Purity specified per lot; commercial grades typically ≥98% (HPLC).
- Supplied as a solid; follow the Certificate of Analysis for storage instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.