Learn More
Selleck Chemical LLC Isradipine 50mg 75695-93-1 PN 200-110
SDP
Supplier:  Selleck Chemical LLC S166250MG
Encompass_Preferred
Isradipine (PN 200-110) is a potent and selective L-type voltage-gated calcium channel blocker, used to treat high blood pressure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-597-3