Learn More
Selleck Chemical LLC ISRIB (trans-isomer) 10mg 1597403-47-8
SDP
Supplier:  Selleck Chemical LLC S740010MG
Encompass_Preferred
ISRIB (trans-isomer), the trans-isomer of ISRIB, is a potent and selective PERK inhibitor with IC50 of 5 nM and does not have global effects on translation, transcription, or mRNA stability in non-stressed cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4740