Learn More
Aobious Inc iST2-1, 200MG
Supplier:  Aobious Inc AOB36416200MG
Encompass
ABIOUS | iST2-1, 2-(4-Methoxyphenyl)-1-((5-(2-nitrophenyl)furan-2-yl)methyl)pyrrolidine | 200MG g | AOB36416-200MG | Purity: 98% by HPLC | 1019158-02-1 | MW: 378.43
Catalog No. NC2888442