Learn More
Cayman Chemical IstradefylIn 10mg
SDP
Supplier:  Cayman Chemical 2295810 MG
Encompass_Preferred
An A2A receptor antagonist (Ki = 2.2 nM in a radioligand binding assay); inhibits haloperidol-induced catalepsy (ED50 = 0.23 mg/kg in rats); alleviates postural defects in a dose-dependent manner without inducing dyskinesias or hyperactivity in an MPTP-induced marmoset model of Parkinson's disease; decreases bradykinesias induced by L-DOPA and improves attentional and working memory deficits in an MPTP-induced macaque model of Parkinson's disease
Catalog No. 50-299-4551