Learn More
Medchemexpress LLC Istradefylline | 155270-99-8 | 99.9% | 384.43 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10888
Encompass_Preferred
Istradefylline is a very potent, selective, and orally active adenosine A2A receptor antagonist, primarily investigated in experimental models of Parkinson's disease. It is intended for research use only.
  • Very potent and selective adenosine A2A receptor antagonist.
  • Orally active compound.
  • Attenuates dopamine and DOPAC depletions in Parkinson's disease models.
  • Reverses drug-induced catalepsy and hypokinesia.
  • Increases locomotor activity and improves motor disability in MPTP-treated marmosets.
  • Shows potentially additive effects with L-DOPA on locomotor activity without increasing dyskinesia.
Catalog No. 50-003-47415