Learn More
Selleck Chemical LLC Isuzinaxib S0777-25MG
SDP
Supplier:  Selleck Chemical LLC S077725MG
Encompass_Preferred
Isuzinaxib (APX-115 free base Ewha-18278 free base) is a potent orally active inhibitor of pan NADPH oxidase (pan-Nox) with Ki of 1 08 M 0 57 M and 0 63 M for Nox1 Nox2 and Nox4 respectively APX-115 free base (Ewha-18278 free base) significantly suppresses the expression of inflammatory molecules including MCP-1/CCL2 IL-6 and TNF in the diabetic kidney
Catalog No. 50-000-42968