Learn More
Medchemexpress LLC IT1t dihydrochloride | 1092776-63-0 | MFCD22683853 | ≥98.0% | 479.57 g/mol | C21H36Cl2N4S2 | 100 MG

Supplier: Medchemexpress LLC HY101458A100MG
IT1t dihydrochloride is a research-grade small-molecule antagonist of the chemokine receptor CXCR4, blocking CXCL12/CXCR4 interaction with an IC50 of 2.1 nM and used in biochemical and cell-based studies including anti-HIV research.
- Potent CXCR4 antagonist (CXCL12/CXCR4 IC50 = 2.1 nM).
- Active against X4-tropic HIV-1 in cell assays (IC50 ≈ 14-19 nM).
- Solid powder suitable for biochemical and cell-based assays.
- Soluble in water (50 mg/mL) and DMSO (≥30 mg/mL) for in vitro use.
- Stable when stored sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
- Molecular weight 479.57 g/mol; formula C21H36Cl2N4S2.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.