Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IT1t dihydrochloride | 1092776-63-0 | MFCD22683853 | 99.8% | 479.57 g/mol | C21H36Cl2N4S2 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY101458A10MM 1ML

Encompass_Preferred

IT1t dihydrochloride is a research-grade small-molecule antagonist of the CXCR4 chemokine receptor. It potently inhibits CXCL12/CXCR4 interaction (IC50 = 2.1 nM) and is used in cellular and animal studies of chemokine signaling and HIV X4 tropism.

  • Potent CXCR4 antagonist with IC50 of 2.1 nM.
  • High purity (~99.8%) suitable for biochemical assays.
  • Soluble in DMSO (≥ 30 mg/mL) and in water (50 mg/mL; ultrasonic recommended).
  • Molecular formula C21H36Cl2N4S2; molecular weight 479.57 g/mol.
  • Recommended storage: sealed at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Reported activity in HIV X4 assays with low-nanomolar IC50 values.

Catalog No. 50-002-49170


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.