Learn More
Medchemexpress LLC Itacnosertib hydrochloride | 2409543-84-4 | 99.43% | 505.01 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY109179A100MG
Encompass_Preferred
Itacnosertib hydrochloride (TP-0184 hydrochloride) is an inhibitor for FLT3, ACVR1 (ALK2, IC50=8 nM), and JAK2 (IC50=8540 nM). It exhibits anti-leukemic activity. It also:
  • Inhibits FLT3, ACVR1 (ALK2), and JAK2.
  • Exhibits anti-leukemic activity.
  • Arrests the cell cycle of MOLM-13, MOLM-14, and MV4-11 at G1/G0 phase.
  • Inhibits the proliferation of acute myeloid leukemia (AML) cells (IC50s for MOLM-13, MOLM-14 and MV4-11 are 6.4, 1.2 and 1.3 nM, respectively).
  • Inhibits activation of FLT3 and ACVR1 downstream signaling pathways.
  • Inhibits serine biosynthesis and amino acid transport pathways.
  • Exhibits antitumor activity against MOLM-13 in mouse models.
Catalog No. 50-003-46299