Learn More
Medchemexpress LLC Itaconic acid | 97-65-4 | MFCD00004260 | 100.0% | 130.10 g/mol | C5H6O4 | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0520R100MG
Encompass_Preferred
Itaconic acid (Standard) is the analytical standard of Itaconic acid This product is intended for research and analytical applications Itaconic acid a precursor of polymers chemicals and fuels can be synthesized by many fungi Itaconic acid also is a macrophage-specific metabolite Itaconic acid mediates crosstalk between macrophage metabolism and peritoneal tumors Itaconic acid has anti-inflammatory anti-microbial and immunomodulatory effect[1][2][4][5]
Catalog No. 50-002-69317