Learn More
Cayman Chemical ITGAMCD11bCTerm RabbIt M 100uL
SDP
Supplier:  Cayman Chemical 32241100 UL
Encompass_Preferred
The nucleoside form of hypoxanthine a product of DNA damage that is formed from 2-deoxyadenosine by adenosine deaminase or RNS enhances 5-FU-induced apoptosis in HT-29 and SW620 cancer cells enhances 5-FU-induced tumor growth suppression in an SW620 mouse xenograft model at 1.6 g/kg
Catalog No. 50-294-9196