Learn More
Aviva Systems Biology Corp ITGB5 Antibody - middle region (ARP45247_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45247_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITGB5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ITGB5. Alternate names: FLJ26658ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58989