Learn More
ABclonal Technology ITIH4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A128071000UL
Encompass_Preferred
The protein encoded by this gene is secreted into the blood, where it is cleaved by plasma kallikrein into two smaller forms. Expression of this gene has been detected only in liver, and it seems to be upregulated during surgical trauma. This gene is part of a cluster of similar genes on chromosome 3. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-15916