Learn More
Medchemexpress LLC Itopride hydrochloride | 122892-31-3 | 99.8% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYB073210MM/1ML
Encompass_Preferred
Itopride hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. It enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, making it useful as a gastrointestinal prokinetic agent. It can be used for researching gastro-esophageal reflux disease (GERD).
  • Potent dopamine-2 antagonist
  • Acetylcholine esterase (AChE) inhibitor
  • Enhances gastric motility through antidopaminergic and anti-acetylcholinesterasic actions
  • Can be used as a gastrointestinal prokinetic agent
  • Useful for researching gastro-esophageal reflux disease (GERD)
Catalog No. 50-004-05371