Learn More
Medchemexpress LLC Itopride hydrochloride | 122892-31-3 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0732100MG
Encompass_Preferred
Itopride hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. It enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, acting as a gastrointestinal prokinetic agent. This chemical can be used for researching gastro-esophageal reflux disease (GERD).
  • Potent dopamine-2 antagonist
  • Acetylcholine esterase (AChE) inhibitor
  • Enhances gastric motility
  • Usable as a gastrointestinal prokinetic agent
  • Used for researching gastro-esophageal reflux disease (GERD)
  • Has prokinetic effects on both the ileum and colon
  • Significantly accelerates peristalsis propagation velocity in ex vivo guinea pig ileum
  • Significantly accelerates gastric emptying in male ddY-strain mice
Catalog No. 50-004-06854