Learn More
Selleck Chemical LLC Itraconazole S2476-200mg
SDP
Supplier:  Selleck Chemical LLC S2476200MG
Encompass_Preferred
Itraconazole is a relatively potent inhibitor of CYP3A4 with IC50 of 6 1 nM used as a triazole antifungal agent Itraconazole is a potent antagonist of the Hedgehog (Hh) signaling pathway Itraconazole suppresses the growth of glioblastoma through induction of autophagy
Catalog No. 50-311-2973