Learn More
Medchemexpress LLC Itraconazole-d9 | 1309272-50-1 | 97.9% | 714.69 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17514S21MG
Encompass_Preferred
Itraconazole-d9 is a deuterium-labeled Itraconazole. It is a triazole antifungal agent and a potent, orally active Hedgehog (Hh) signaling pathway antagonist with an IC50 of approximately 800 nM. It inhibits lanosterol 14α-demethylase (cytochrome P450 enzyme), thereby blocking the oxidative conversion of lanosterol to ergosterol. Itraconazole also exhibits anticancer and antiangiogenic effects and acts as an oxysterol-binding protein (OSBP) inhibitor.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-86312