Learn More
Medchemexpress LLC IU1-248 | 2307472-03-1 | 99.2% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12288525MG
Encompass_Preferred
IU1-248 is a small-molecule derivative that acts as a potent and selective inhibitor of the deubiquitinase USP14, reported to have an IC50 of 0.83 μM.
  • Potent and selective USP14 inhibitor; reported IC50 0.83 μM.
  • High purity (≈99.2%), suitable for biochemical and cell-based research.
  • Supplied as a solid with multiple pack sizes, including 25 MG for small-scale studies.
  • Available as DMSO solutions for convenient dosing in assays.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-32624