Learn More
Medchemexpress LLC IU1-248 | 2307472-03-1 | 99.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1228855MG
Encompass_Preferred
IU1-248 is a derivative of IU1 and a potent, selective inhibitor of the deubiquitinase USP14 (IC50 = 0.83 μM). It is supplied as a solid for research use and is commonly used in biochemical and cell-based studies targeting ubiquitin-proteasome system pathways.
  • Potent USP14 inhibitor with IC50 = 0.83 μM.
  • Selective activity suitable for biochemical assays and cell studies.
  • Available in small laboratory quantities for research applications.
  • Stable when stored as a powder at recommended temperatures.
Catalog No. 50-002-31658