Learn More
Medchemexpress LLC IU1-248 | 2307472-03-1 | 99.2% | 50 MG

Supplier: Medchemexpress LLC HY12288550MG
IU1-248 is a derivative of IU1 and a potent, selective inhibitor of the deubiquitinating enzyme USP14 (IC50 = 0.83 μM). It is supplied as a solid with high reported purity for research use.
- Potent, selective USP14 inhibitor (IC50 = 0.83 μM).
- High purity, 99.19% by HPLC.
- Molecular formula C20H23N3O2; molecular weight 337.42 g·mol⁻1.
- Appearance: solid, light brown to khaki.
- Storage: powder long-term at -20°C; in solvent store at -80°C for up to 6 months.
- Available in multiple pack sizes including 50 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.