Learn More
Takara Bio PrimeScript™ IV 1st strand cDNA Synthesis Mix 50 Rxns
SDP
Supplier:  Takara Bio 6215A
Encompass
PrimeScript IV 1st strand cDNA Synthesis Mix contains a 5X premix for synthesizing first-strand cDNA from total RNA or poly A+ RNA. A cDNA synthesis reaction can be started simply by adding template RNA and water to the premix, which already contains PrimeScript IV Reverse Transcriptase, RNase Inhibitor, Oligo dT Primer, dNTP Mixture, and reaction buffer. The premix also contains an accessory protein that eliminates the need for RNA denaturation, which is commonly performed before reverse transcription.
Catalog No. NC2741027