Learn More
Med Vet International IV Fluid Hanger w/Pulley System, J0400S, Each
Supplier:  Med Vet International JOR400S
Encompass_Preferred
IV fluid Hanger with Pulley System.This handy system allows easier handling of heavy and large bags of IV fluids.  The fluid can be raised and lowered as needed. The hanger swivels 360 degrees to avoid tangling line. Use in conjunction with J0400LN coiled IV set.   J0400s Complete System (as shown) J0400r Individual Hanger Only
Catalog No. 50-005-85294