Learn More
Health Care Logistics Inc IV INSPECTION LIGHT BOX 1/EA
SDP
Supplier:  Health Care Logistics Inc 17109
Encompass
•Determine the absence or inclusion of particulates in compounded sterile drug products and perform finished preparation release checks for purity and quality. •Physical inspection of the compounded sterile product (CSP) is achieved against lighted White and Black backgrounds. •4" Dia., 2X magnifying glass allows for close visual inspection. Flexible arm permits easy movement. •Consists of one Black UV and one White LED light. Both 15 watt lights are included and a 5' cord. •One 110V wall outlet is needed for operation. Both lights are independently operated. •Predrilled mounting holes. •Dimensions: 19-3/4"W x 24-1/2"H x 7"D
Catalog No. NC0076965