Learn More
Medicus Health Iv Pole Clamp For Poles Up To 2" In Diameter
SDP
Supplier:  Medicus Health 2842M1
Encompass
∙ Aluminum Pole Clamp is used to mount most accessories to IV Poles without disassembling pole & it attaches to IV Poles up to 2" in diameter ∙Attaches to IV Poles up to 2" in diameter∙Easy position adjustments∙Set-screw locks accessories in place
Catalog No. NC1728846