Learn More
Apexbio Technology LLC ACPT-I 194918-76-8 10mg
SDP
Supplier:  Apexbio Technology LLC B505110
Encompass_Preferred
ACPT-I (CAS 194918-76-8) is a small molecule agonist that selectively targets group III metabotropic glutamate receptors (mGluRs) By modulating glutamatergic neurotransmission ACPT-I has been shown in preclinical studies to confer neuroprotection reduce central nervous system excitability and exhibit anticonvulsant as well as anxiolytic properties These pharmacological effects support its use in research on neurodegenerative disorders and epilepsy where it serves as a tool to elucidate the role of group III mGluRs in neuronal survival and excitability regulation
Catalog No. 50-000-88714