Learn More
Selleck Chemical LLC Ivabradine HCl S2086-10mg
SDP
Supplier:  Selleck Chemical LLC S208610MG
Encompass_Preferred
Ivabradine HCl (S 16257-2 S-18982 D6 hydrochloride) a new If inhibitor with IC 50 of 2 9 M which acts specifically on the pacemaker activity of the sinoatrial node is a pure heart rate lowering agent
Catalog No. 50-311-1266