Learn More
TARGETMOL CHEMICALS INC IVACAFTOR 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T258810MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ivacaftor (VX-770) (VX-770) is a potentiator of CFTR targeting G551D-CFTR (EC50 100 nM) and F508del-CFTR (EC50 25 nM) in Fisher rat thyroid cells respectively. purity: 99%
Catalog No. 50-257-7088