Learn More
Selleck Chemical LLC Ivacaftor (VX-770) S1144-200mg
SDP
Supplier:  Selleck Chemical LLC S1144200MG
Encompass_Preferred
Ivacaftor (VX-770) is a selective potentiator of CFTR targeting G551D-CFTR and F508del-CFTR with EC50 of 100 nM and 25 nM in fisher rat thyroid cells respectively
Catalog No. 50-311-1472