Learn More
Medchemexpress LLC Ivacaftor | 873054-44-5 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13017100MG
Encompass_Preferred
Ivacaftor is a potent and orally bioavailable compound that functions as a CFTR potentiator. It is effective in targeting both G551D-CFTR and F508del-CFTR mutations, demonstrating EC50s of 100 nM and 25 nM respectively. This product is intended for research use only.
  • Potent CFTR potentiator
  • Orally bioavailable
  • Targets G551D-CFTR
  • Targets F508del-CFTR
  • For research use only
Catalog No. 50-003-39042