Learn More
Medchemexpress LLC Ivacaftor-d9 | 1413431-07-8 | 99.8% | C24H19D9N2O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13017S10MG
Encompass_Preferred
Ivacaftor-d9 is a deuterium-labeled isotopologue of ivacaftor used as an analytical reference and internal standard for LC-MS/MS, pharmacokinetic, and ADME studies. The nine deuterium atoms enable unambiguous mass separation from non-labeled ivacaftor, improving assay specificity and quantitation.
  • Deuterated internal standard for LC-MS/MS assays.
  • Enables accurate quantification and reduced matrix effects.
  • Suitable for pharmacokinetic and ADME studies.
  • High chemical purity (99.8%).
  • Stable under typical laboratory storage conditions.
Catalog No. 50-002-60167