Learn More
Medchemexpress LLC Ivermectin B1a | 71827-03-7 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12693710MM/1ML
Encompass_Preferred
Ivermectin B1a is a main component of Ivermectin and a derivative of Avermectin B1a. It functions as a broad-spectrum anti-parasite agent and is a candidate therapeutic against SARS-CoV-2/COVID-19. This compound belongs to the macrocyclic lactone class of avermectins.
  • Functions as a broad-spectrum anti-parasite agent.
  • Main component of Ivermectin.
  • Identified as a candidate therapeutic against SARS-CoV-2/COVID-19.
  • Belongs to the macrocyclic lactone class of avermectins.
  • Purity of 99.97%.
  • Available as a 10 mM solution in DMSO.
Catalog No. 50-003-42196