Learn More
Medchemexpress LLC Ivermectin B1a | 71827-03-7 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12693710MG
Encompass_Preferred
Ivermectin B1a is a derivative of Avermectin B1a and a main component of Ivermectin. Ivermectin is a broad-spectrum anti-parasite agent and a candidate therapeutic against SARS-CoV-2/COVID-19. It belongs to the macrocyclic lactone class of avermectins, consisting of a mixture of ivermectin B1a (not less than 80%) and ivermectin B1b (not more than 20%).
  • Derivative of Avermectin B1a
  • Main component of Ivermectin
  • Broad-spectrum anti-parasite agent
  • Candidate therapeutic against SARS-CoV-2/COVID-19
  • Belongs to macrocyclic lactone class of avermectins
Catalog No. 50-003-41338