Learn More
Medchemexpress LLC Ivermectin B1b | 70209-81-3 | MFCD00866559 | 98.7% | 861.07 | C47H72O14 | 1MG

Supplier: Medchemexpress LLC HY1257291MG
Ivermectin B1b is the minor component of ivermectin, a macrocyclic lactone antiparasitic agent supplied for research use. The compound has documented analytical data and literature reports of antiviral activity in cell culture; product listings include CAS, formula, molecular weight, and RP-HPLC documentation.
- Minor component of ivermectin with antiparasitic activity.
- Macrocyclic lactone structure suitable for research applications.
- Reported to inhibit replication of SARS-CoV-2 in cell culture.
- CAS number 70209-81-3 for unambiguous identification.
- Chemical formula C47H72O14; molecular weight ≈ 861.07.
- High purity reported (about 98.66%) with RP-HPLC documentation available.
- Available in small laboratory quantities (500 μg-10 mg) for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.