Learn More
Medchemexpress LLC Ivermectin-d2 | 71827-03-7 | 97.6% | 877.11 | C48H72D2O14 | 1 MG

Supplier: Medchemexpress LLC HYW6542441MG
Ivermectin-d2 is a deuterium-labeled analogue of ivermectin supplied as a research-grade standard for analytical and research applications. It is provided as a solid, with specified purity and storage conditions to support handling and long-term stability.
- Deuterium-labeled analogue of ivermectin suitable for analytical use.
- Provided as a solid, white to light yellow.
- Purity 97.6% (rounded from manufacturer value).
- Molecular formula C48H72D2O14; molecular weight 877.11.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Intended for research and analytical applications only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.