Learn More
CORETEX MANUFACTURE Ivy X Post-Contact Poison Oak & Ivy Cleanser, 2oz Bottle
Supplier:  CORETEX MANUFACTURE 84332
Encompass_Preferred
Standard soap and water will not emulsify poisonous plant oils. Ivy X Post-ContactSkin Cleanser is designed to be “Wetter than Water” to start the emulsifying processto assist in removing plant oils before they have time to cause the uncomfortablerash, blisters and itching commonly associated with poisonous plants. Ourpackaging (towelettes and bottles) allows for a compliant and affordable skinprotection program to help in poisonous plant protection.
Catalog No. NC1647475