Learn More
Apexbio Technology LLC IWP-2, 50mg. Cas: 686770-61-6 MFCD: MFCD04457599. Wnt production inhibitor,PORCN inhibitor
SDP
Supplier:  Apexbio Technology LLC A351250
Encompass_Preferred
IC50: 27 nM for Wnt pathway activityThe Wnt signaling pathway is important in embryonic development as well as the initiation and progression of a number of types of human cancer. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4808