Learn More
Medchemexpress LLC HY-13912 5mg , IWP-2 CAS:686770-61-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY139125MG
Encompass_Preferred
HY-13912 5mg IWP-2 CAS: 686770-61-6 IWP-2 is an inhibitor of Wnt processing and secretion with an IC50 of 27 nM. IWP-2 targets the membrane-bound O-acyltransferase porcupine (Porcn) and thus preventing a crucial Wnt ligand palmitoylation. IWP-2 is also an ATP-competitive CK1δ inhibitor with an IC50 of 40 nM for the gatekeeper mutant M82FCK1δ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0115