Learn More
STEMCELL Technologies IWP-3, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72544
Encompass
Inhibitor of WNT Production-3 (IWP-3) inhibits WNT signaling by inactivating Porcupine,  a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.).  IWP-3 impairs WNT pathway activity in vitro with an IC₅₀ value of 40 nM (Chen et al.). DIFFERENTIATION· Promotes cardiomyocyte differentiation from human embryonic stem cells that have been induced to mesoderm by addition of BMP4 and Activin A (Willems et al.).
Catalog No. 50-197-5940