Learn More
Cayman Chemical IWR-1-endo, 1127442-82-3, 5 mg
SDP
Supplier:  Cayman Chemical 136595
Encompass_Preferred
A potent inhibitor of the Wnt response, blocking a cell-based Wnt/β-catenin pathway reporter response (IC50 = 180 nM); inhibits Wnt-induced accumulation of β-catenin, leading to proteasomal degradation of this protein
Catalog No. NC1432243