Learn More
Medchemexpress LLC HY-10452 5mg , Ixazomib citrate CAS:1239908-20-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104525MG
Encompass_Preferred
HY-10452 5mg Ixazomib citrate CAS: 1239908-20-3 Ixazomib citrate (MLN9708) is a reversible inhibitor of the chymotrypsin-like proteolytic β5 site of the 20S proteasome with an IC50 of 3.4 nM and a Ki of 0.93 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0467