Learn More
Selleck Chemical LLC Ixazomib (MLN2238) 5mg 1072833-77-2
SDP
Supplier:  Selleck Chemical LLC S21805MG
Encompass_Preferred
Ixazomib (MLN2238) inhibits the chymotrypsin-like proteolytic (?5) site of the 20S proteasome with IC50 and Ki of 3.4 nM and 0.93 nM in cell-free assays, respectively, also inhibits the caspase-like (?1) and trypsin-like (?2) proteolytic sites, with IC50 of 31 and 3500 nM. Ixazomib (MLN2238) induces autophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2137