Learn More
Medchemexpress LLC Izuforant | 1429374-83-3 | C12H12BrN7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14727610MG
Encompass_Preferred
Izuforant is a small-molecule research compound that acts as a selective histamine H4 receptor antagonist. It is orally active and has demonstrated potent activity versus human H4R (IC50 = 36 nM), with reported anti-pruritic and anti-inflammatory effects; the compound also shows weaker binding to the 5-HT3 receptor (IC50 ≈ 9.1 μM). Izuforant is provided as a research standard for laboratory and preclinical studies.
  • Potent H4R antagonist activity (IC50 = 36 nM).
  • Demonstrates anti-pruritic and anti-inflammatory effects.
  • Orally active small-molecule suitable for in vivo studies.
  • Secondary 5-HT3 receptor binding at micromolar potency.
  • Molecular formula C12H12BrN7; molecular weight 334.17.
  • Supplied as a high-purity research standard for laboratory use.
Catalog No. 50-002-59524